2-Bromo-4-methoxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4-methoxybenzeneboronic acid

27831-1G 27831-25G 27831-500MG 27831-5G
76900-822EA 260.99 USD
76900-822 76900-824 76900-826 76900-828
2-Bromo-4-methoxyphenylboronic acid ≥97%
2-Bromo-4-methoxyphenylboronic acid
Formula: C₇H₈BBrO₃
MW: 230.852 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28142623
CAS Number: 1879166-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR