(2-Chloro-3-fluoropyridin-4-yl)methanol ≥97% research grade

Supplier: AOBChem USA

Warning

27786-1G 27786-25G 27786-5G
76900-810EA 64.72 USD
76900-810 76900-812 76900-814
(2-Chloro-3-fluoropyridin-4-yl)methanol ≥97% research grade
(2-Chloro-3-fluoropyridin-4-yl)methanol
Formula: C₆H₅ClFNO
Storage Temperature: Refrigerator
MDL Number: MFCD11045047
CAS Number: 946127-54-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR