2-Bromo-5-chloro-4-methoxyacetophenone ≥97%
Supplier: AOBChem USA
|
Warning
|
27767-25G
27767-5G
76900-798EA
310.85
USD
76900-798
76900-800
2-Bromo-5-chloro-4-methoxyacetophenone ≥97%
2-Bromo-5-chloro-4-methoxyacetophenone
Formula:
C₉H₈BrClO₂ Storage Temperature: Ambient |
MDL Number:
MFCD28142619 CAS Number: 1809168-61-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...