2-Bromo-3-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

27742-1G 27742-25G 27742-5G
76900-784EA 111.4 USD
76900-784 76900-786 76900-788
2-Bromo-3-(trifluoromethyl)phenol ≥97%
2-Bromo-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄BrF₃O
MW: 241.005 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD13194487
CAS Number: 1214323-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR