2-Bromo-5-methoxybenzamide ≥97%

Supplier: AOBChem USA

Warning

27611-25G 27611-5G
76900-744EA 187.78 USD
76900-744 76900-746
2-Bromo-5-methoxybenzamide ≥97%
2-Bromo-5-methoxybenzamide

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
Storage Temperature: Ambient
MDL Number: MFCD00208336
CAS Number: 99848-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR