2-Chloro-3-methoxypyridine-4-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

27587-1G 27587-25G 27587-500MG 27587-5G
76900-728EA 203.7 USD
76900-728 76900-730 76900-732 76900-734
2-Chloro-3-methoxypyridine-4-carbaldehyde ≥97%
2-Chloro-3-methoxypyridine-4-carbaldehyde
Formula: C₇H₆ClNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD16606537
CAS Number: 905563-05-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR