2-Bromo-6-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

27482-1G 27482-25G 27482-500MG 27482-5G
76900-702EA 163.38 USD
76900-702 76900-704 76900-706 76900-708
2-Bromo-6-methoxybenzyl alcohol ≥97%
2-Bromo-6-methoxybenzyl alcohol
Formula: C₈H₉BrO₂
MW: 217.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18391885
CAS Number: 93710-52-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR