2-Fluoro-4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%

Supplier: AOBChem USA

Warning

27390-1G 27390-25G 27390-500MG 27390-5G
76900-684EA 199.45 USD
76900-684 76900-686 76900-688 76900-690
2-Fluoro-4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥97%
2-Fluoro-4-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Formula: C₁₂H₁₇BFNO₂
MW: 237.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16996672
CAS Number: 1363192-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR