2-Hydroxy-4-methylphenylboronic acid pinacol ester ≥97% research grade

Supplier: AOBChem USA

Warning

27328-1G 27328-500MG 27328-5G
76900-672EA 498.63 USD
76900-672 76900-674 76900-676
2-Hydroxy-4-methylphenylboronic acid pinacol ester ≥97% research grade
2-Hydroxy-4-methylphenylboronic acid pinacol ester
Formula: C₁₃H₁₉BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD16994281
CAS Number: 517864-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR