2-Bromo-4-fluoro-6-iodoaniline ≥95%

Supplier: AOBChem USA

Warning

27265-1G 27265-25G 27265-500MG 27265-5G
76900-638EA 388.29 USD
76900-638 76900-640 76900-642 76900-644
2-Bromo-4-fluoro-6-iodoaniline ≥95%
2-Bromo-4-fluoro-6-iodoaniline

Precaución: for Research Use Only

Formula: C₆H₄BrFIN
Storage Temperature: Ambient
MDL Number: MFCD24160442
CAS Number: 1447606-71-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR