2-Methyl-5-phenylpyridine ≥95%

Supplier: AOBChem USA

Warning

27190-1G 27190-25G 27190-5G
76900-618EA 67.9 USD
76900-618 76900-620 76900-622
2-Methyl-5-phenylpyridine ≥95%
2-Methyl-5-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
Storage Temperature: Refrigerator
CAS Number: 3256-88-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR