2-Fluoro-4-methoxybenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

27162-25G 27162-5G
76900-608EA 559.1 USD
76900-608 76900-610
2-Fluoro-4-methoxybenzyl alcohol ≥97%
2-Fluoro-4-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MW: 156.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04116336
CAS Number: 405-09-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR