2-Bromo-1-fluoro-3-iodobenzene ≥97%

Supplier: AOBChem USA

Warning

27119-25G 27119-5G
76900-598EA 421.18 USD
76900-598 76900-600
2-Bromo-1-fluoro-3-iodobenzene ≥97%
2-Bromo-1-fluoro-3-iodobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrFI
MW: 300.8949 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09056796
CAS Number: 851368-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR