2-Fluoro-3-(trifluoromethyl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-3-hydroxybenzotrifluoride

27098-25G 27098-5G
76900-592EA 346.92 USD
76900-592 76900-594
2-Fluoro-3-(trifluoromethyl)phenol ≥97%
2-Fluoro-3-(trifluoromethyl)phenol

Precaución: for Research Use Only

Formula: C₇H₄F₄O
MW: 180.1 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00083554
CAS Number: 207291-85-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR