2-Cyanopyridine-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (6-Cyanopyridin-3-yl)boronic acid

27018-1G 27018-25G 27018-5G
76900-556EA 62.6 USD
76900-556 76900-558 76900-560
2-Cyanopyridine-5-boronic acid ≥97%
2-Cyanopyridine-5-boronic acid

Precaución: for Research Use Only

Formula: C₆H₅BN₂O₂
MW: 147.927 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10696633
CAS Number: 1011722-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR