2-Methoxy-4-methylphenylboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-(2-Methoxy-4-methylphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

26910-1G 26910-25G 26910-500MG 26910-5G
76900-532EA 290.69 USD
76900-532 76900-534 76900-536 76900-538
2-Methoxy-4-methylphenylboronic acid pinacol ester ≥97%
2-Methoxy-4-methylphenylboronic acid pinacol ester
Formula: C₁₄H₂₁BO₃
MW: 248.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18732610
CAS Number: 1326662-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR