2-Amino-5-(4-trifluoromethoxyphenyl)pyridine ≥97%

Supplier: AOBChem USA

Warning

26893-1G 26893-25G 26893-5G
76900-518EA 97.61 USD
76900-518 76900-520 76900-522
2-Amino-5-(4-trifluoromethoxyphenyl)pyridine ≥97%
2-Amino-5-(4-trifluoromethoxyphenyl)pyridine
Formula: C₁₂H₉F₃N₂O
MW: 254.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD17496170
CAS Number: 1110656-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR