2,6-Dibromo-p-xylene ≥95%

Supplier: AOBChem USA

Warning

26835-1G 26835-25G 26835-500MG 26835-5G
76900-502EA 147.47 USD
76900-502 76900-504 76900-506 76900-508
2,6-Dibromo-p-xylene ≥95%
2,6-Dibromo-p-xylene
Formula: C₈H₈Br₂
MW: 263.957 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11845880
CAS Number: 66788-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR