2-Chloro-4-fluoro-3-(methoxymethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

26804-1G 26804-25G 26804-500MG 26804-5G
76900-490EA 280.08 USD
76900-490 76900-492 76900-494 76900-496
2-Chloro-4-fluoro-3-(methoxymethyl)benzoic acid ≥97%
2-Chloro-4-fluoro-3-(methoxymethyl)benzoic acid
Formula: C₉H₈ClFO₃
MW: 218.609 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32631796
CAS Number: 2404733-74-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR