2-Bromo-6-methoxy-4-nitroaniline ≥97%

Supplier: AOBChem USA

Warning

26709-25G 26709-5G
76900-434EA 488.02 USD
76900-434 76900-436
2-Bromo-6-methoxy-4-nitroaniline ≥97%
2-Bromo-6-methoxy-4-nitroaniline

Precaución: for Research Use Only

Formula: C₇H₇BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD05863343
CAS Number: 16618-66-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR