2,6-Dichloro-4-hydroxypyridine ≥97%

Supplier: AOBChem USA

Warning

26704-25G 26704-5G
76900-422EA 516.66 USD
76900-422 76900-424
2,6-Dichloro-4-hydroxypyridine ≥97%
2,6-Dichloro-4-hydroxypyridine
Formula: C₅H₃Cl₂NO
Storage Temperature: Ambient
MDL Number: MFCD00955619
CAS Number: 17228-74-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR