2-(Hydroxymethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Hydroxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzenemethanol

26668-1G 26668-25G 26668-500MG 26668-5G
76900-410EA 224.92 USD
76900-410 76900-412 76900-414 76900-416
2-(Hydroxymethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%
2-(Hydroxymethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₉BO₄
MW: 250.099 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18729969
CAS Number: 760989-96-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR