2,6-Dichloro-4-fluorobenzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

26324-1G 26324-25G 26324-5G
76900-348EA 90.18 USD
76900-348 76900-350 76900-352
2,6-Dichloro-4-fluorobenzyl alcohol ≥97%
2,6-Dichloro-4-fluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅Cl₂FO
MW: 195.018 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD28133674
CAS Number: 1806353-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR