4-Chloro-2-fluoro-1,1'-biphenyl ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-4-chloro biphenyl

26277-1G 26277-25G 26277-500MG 26277-5G
76900-330EA 224.92 USD
76900-330 76900-332 76900-334 76900-336
4-Chloro-2-fluoro-1,1'-biphenyl ≥97%
4-Chloro-2-fluoro-1,1'-biphenyl
Formula: C₁₂H₈ClF
MW: 206.64 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD20267653
CAS Number: 39224-18-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR