2-Chloro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%

Supplier: AOBChem USA

Warning

26231-1G 26231-25G 26231-500MG 26231-5G
76900-314EA 260.99 USD
76900-314 76900-316 76900-318 76900-320
2-Chloro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97%
2-Chloro-6-methyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Formula: C₁₃H₁₈BClO₃
MW: 268.54 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22414580
CAS Number: 1294518-25-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR