2,6-Dichloro-4-fluorobenzoic acid ≥97%

Supplier: AOBChem USA

Warning

26201-1G 26201-25G 26201-500MG 26201-5G
76900-300EA 290.69 USD
76900-300 76900-302 76900-304 76900-306
2,6-Dichloro-4-fluorobenzoic acid ≥97%
2,6-Dichloro-4-fluorobenzoic acid
Formula: C₇H₃Cl₂FO₂
MW: 209.00192 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23135660
CAS Number: 232275-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR