6-Fluoro-3-iodo-2-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-iodo-6-methylpyridine, 2-Fluoro-5-iodo-6-picoline, 6-Fluoro-3-iodo-2-picoline

26176-1G 26176-25G 26176-5G
76900-270EA 62.6 USD
76900-270 76900-272 76900-274
6-Fluoro-3-iodo-2-methylpyridine ≥97%
6-Fluoro-3-iodo-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅FIN
MW: 237.02 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD08277264
CAS Number: 884495-23-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR