2-Chloro-3-(trifluoromethyl)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

26167-25G 26167-5G
76900-262EA 190.97 USD
76900-262 76900-264
2-Chloro-3-(trifluoromethyl)benzyl alcohol ≥97%
2-Chloro-3-(trifluoromethyl)benzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₆ClF₃O
Storage Temperature: Ambient
MDL Number: MFCD01631537
CAS Number: 261763-20-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR