4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-hydroxyphenylboronic acid, pinacol ester

26085-1G 26085-25G 26085-5G
76900-224EA 101.85 USD
76900-224 76900-226 76900-228
4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol ≥97% research grade
4-Fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Formula: C₁₂H₁₆BFO₃
MW: 238.06 g/mol
Storage Temperature: Ambient
MDL Number: MFCD16994432
CAS Number: 1398923-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR