2-Iodo-4-(trifluoromethoxy)phenol ≥97%

Supplier: AOBChem USA

Warning

26061-25G 26061-5G
76900-216EA 453.01 USD
76900-216 76900-218
2-Iodo-4-(trifluoromethoxy)phenol ≥97%
2-Iodo-4-(trifluoromethoxy)phenol

Precaución: for Research Use Only

Formula: C₇H₄F₃IO₂
Storage Temperature: Ambient
CAS Number: 405517-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR