2-Bromo-5-iodophenol ≥97%

Supplier: AOBChem USA

Warning

26006-1G 26006-25G 26006-5G
76900-196EA 76.39 USD
76900-196 76900-198 76900-200
2-Bromo-5-iodophenol ≥97%
2-Bromo-5-iodophenol

Precaución: for Research Use Only

Formula: C₆H₄BrIO
Storage Temperature: Refrigerator
MDL Number: MFCD09907878
CAS Number: 932372-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR