2-Phenylthiazole ≥95%

Supplier: AOBChem USA

Warning

25896-25G 25896-5G
76900-158EA 233.4 USD
76900-158 76900-160
2-Phenylthiazole ≥95%
2-Phenylthiazole

Precaución: for Research Use Only

Formula: C₉H₇NS
MW: 161.2200 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00272327
CAS Number: 1826-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR