2-Methoxy-4-biphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

25872-1G 25872-500MG 25872-5G
76900-138EA 524.09 USD
76900-138 76900-140 76900-142
2-Methoxy-4-biphenylboronic acid ≥97%
2-Methoxy-4-biphenylboronic acid
Formula: C₁₃H₁₃BO₃
Storage Temperature: Refrigerator
MDL Number: MFCD28806016
CAS Number: 2121511-97-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR