2-Chloro-4-fluoro-5-iodopyridine ≥97% research grade

Supplier: AOBChem USA

Warning

25753-1G 25753-25G 25753-500MG 25753-5G
76900-100EA 203.7 USD
76900-100 76900-102 76900-104 76900-106
2-Chloro-4-fluoro-5-iodopyridine ≥97% research grade
2-Chloro-4-fluoro-5-iodopyridine
Formula: C₅H₂ClFIN
Storage Temperature: Ambient
MDL Number: MFCD20040082
CAS Number: 1370534-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR