2,5-Dibromo-3-methoxypyridine ≥97%

Supplier: AOBChem USA

Warning

25728-1G 25728-25G 25728-500MG 25728-5G
76900-078EA 141.1 USD
76900-078 76900-080 76900-082 76900-084
2,5-Dibromo-3-methoxypyridine ≥97%
2,5-Dibromo-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅Br₂NO
Storage Temperature: Refrigerator
CAS Number: 1142191-57-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR