2,6-Difluoro-4-(methylthio)benzyl alcohol ≥97%

Supplier: AOBChem USA

Warning

25564-1G 25564-25G 25564-500MG 25564-5G
76900-024EA 260.99 USD
76900-024 76900-026 76900-028 76900-030
2,6-Difluoro-4-(methylthio)benzyl alcohol ≥97%
2,6-Difluoro-4-(methylthio)benzyl alcohol
Formula: C₈H₈F₂OS
Storage Temperature: Refrigerator
MDL Number: MFCD23713130
CAS Number: 1428234-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR