2-Fluoro-4-(methylthio)pyridine ≥97% research grade

Supplier: AOBChem USA

Warning

25523-1G 25523-25G 25523-500MG 25523-5G
76900-012EA 379.81 USD
76900-012 76900-014 76900-016 76900-018
2-Fluoro-4-(methylthio)pyridine ≥97% research grade
2-Fluoro-4-(methylthio)pyridine
Formula: C₆H₆FNS
MW: 143.182 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23713127
CAS Number: 1428234-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR