2,6-Difluoro-4-(methylthio)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

25498-1G 25498-25G 25498-500MG 25498-5G
76899-996EA 290.69 USD
76899-996 76899-998 76900-000 76900-002
2,6-Difluoro-4-(methylthio)phenylboronic acid ≥97%
2,6-Difluoro-4-(methylthio)phenylboronic acid
Formula: C₇H₇BF₂O₂S
Storage Temperature: Refrigerator
MDL Number: MFCD23713121
CAS Number: 1451392-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR