2,6-Difluoro-3-methoxyphenol ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,4-Difluor-3-hydroxyanisole

25457-1G 25457-25G 25457-5G
76899-988EA 79.57 USD
76899-988 76899-990 76899-992
2,6-Difluoro-3-methoxyphenol ≥97%
2,6-Difluoro-3-methoxyphenol

Precaución: for Research Use Only

Formula: C₇H₆F₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04115907
CAS Number: 886498-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR