2-Amino-5-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

25430-25G 25430-5G
76899-970EA 240.83 USD
76899-970 76899-972
2-Amino-5-phenylpyridine ≥97%
2-Amino-5-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₀N₂
Storage Temperature: Ambient
MDL Number: MFCD01692452
CAS Number: 33421-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR