2-Methoxy-5-(trifluoromethyl)benzoic acid ≥97%

Supplier: AOBChem USA

Warning

25405-25G 25405-5G
76899-962EA 240.83 USD
76899-962 76899-964
2-Methoxy-5-(trifluoromethyl)benzoic acid ≥97%
2-Methoxy-5-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇F₃O₃
Storage Temperature: Ambient
MDL Number: MFCD01862091
CAS Number: 4864-01-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR