2,6-Dichloro-4-iodoaniline ≥97%
Supplier: AOBChem USA
|
Warning
|
25285-1G
25285-25G
25285-5G
76899-952EA
117.76
USD
76899-952
76899-954
76899-956
2,6-Dichloro-4-iodoaniline ≥97%
2,6-Dichloro-4-iodoaniline
Formula:
C₆H₄Cl₂IN MW: 287.913 g/mol Storage Temperature: Ambient |
CAS Number:
697-89-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...