2,5-Difluorobenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Difluorophenylboronic acid pinacol ester, 2-(2,5-Difluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

25259-25G 25259-5G
76899-940EA 310.85 USD
76899-940 76899-942
2,5-Difluorobenzeneboronic acid pinacol ester ≥97%
2,5-Difluorobenzeneboronic acid pinacol ester
Formula: C₁₂H₁₅BF₂O₂
MW: 240.057 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12405346
CAS Number: 408492-25-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR