2-Bromo-4-(methoxymethyl)fluorobenzene ≥95%

Supplier: AOBChem USA

Warning

25252-1G 25252-25G 25252-5G
76899-926EA 64.72 USD
76899-926 76899-928 76899-930
2-Bromo-4-(methoxymethyl)fluorobenzene ≥95%
2-Bromo-4-(methoxymethyl)fluorobenzene

Precaución: for Research Use Only

Formula: C₈H₈BrFO
Storage Temperature: Refrigerator
MDL Number: MFCD04116324
CAS Number: 887268-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR