2,6-Dichloro-4-fluoroiodobenzene ≥97%

Supplier: AOBChem USA

Warning

25239-25G 25239-5G
76899-916EA 396.78 USD
76899-916 76899-918
2,6-Dichloro-4-fluoroiodobenzene ≥97%
2,6-Dichloro-4-fluoroiodobenzene

Precaución: for Research Use Only

Formula: C₆H₂Cl₂FI
Storage Temperature: Refrigerator
MDL Number: MFCD13194304
CAS Number: 939990-10-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR