2-Chloro-3-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

25208-1G 25208-25G 25208-500MG 25208-5G
76899-900EA 213.25 USD
76899-900 76899-902 76899-904 76899-906
2-Chloro-3-phenylpyridine ≥97%
2-Chloro-3-phenylpyridine
Formula: C₁₁H₈ClN
MW: 189.641 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114133
CAS Number: 31557-57-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR