2-Fluoro-4-phenylpyridine ≥97%

Supplier: AOBChem USA

Warning

25176-1G 25176-25G 25176-500MG 25176-5G
76899-888EA 231.28 USD
76899-888 76899-890 76899-892 76899-894
2-Fluoro-4-phenylpyridine ≥97%
2-Fluoro-4-phenylpyridine
Formula: C₁₁H₈FN
MW: 173.186 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04114171
CAS Number: 116241-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR