2-Chloro-3,5-dibromobenzotrifluoride ≥95%

Supplier: AOBChem USA

Warning

25160-1G 25160-25G 25160-5G
76899-878EA 84.88 USD
76899-878 76899-880 76899-882
2-Chloro-3,5-dibromobenzotrifluoride ≥95%
2-Chloro-3,5-dibromobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂Br₂ClF₃
MW: 338.3500 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09800690
CAS Number: 1027512-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR