2-(2-Chloro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-4-(trifluoromethyl)phenylboronic acid pinacol ester

25117-1G 25117-25G 25117-5G
76899-868EA 62.6 USD
76899-868 76899-870 76899-872
2-(2-Chloro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97% research grade
2-(2-Chloro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₃H₁₅BClF₃O₂
MW: 306.51 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18756701
CAS Number: 1165935-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR