2-Bromo-1,3-dimethyl-5-fluorobenzene ≥97%

Supplier: AOBChem USA

Warning

25043-25G 25043-5G
76899-820EA 177.18 USD
76899-820 76899-822
2-Bromo-1,3-dimethyl-5-fluorobenzene ≥97%
2-Bromo-1,3-dimethyl-5-fluorobenzene
Formula: C₈H₈BrF
Storage Temperature: Ambient
MDL Number: MFCD09864702
CAS Number: 14659-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR